Learn More
Biotium Ethidium Homodimer I
Catalog No. 501964624
Encompass_Preferred
Ethidium Homodimer I (EthD-1) is a high-affinity fluorescent nucleic acid stain. It binds to both DNA and RNA in a sequence-independent manner and with a >30-fold fluorescence enhancement. It is useful for detecting nucleic acids in solution, or for selectively staining dead cells with damaged plasma membranes. Biotium offers Ethidium Homodimer I in much higher purity than other manufacturers. The dye has an excitation/emission of 528/617 nm when bound to DNA. Properties: Red solid soluble in water or DMSO; Store dessicated at 4°C, protect from light; Ex/Em = 528/617 nm (with DNA); C46H50Cl4N8; MW:857; [61926-22-5].
Catalog No. 50-196-4624 Supplier Biotium Supplier No. 40010
Ethidium Homodimer I (EthD-1) is a high-affinity fluorescent nucleic acid stain. It binds to both DNA and RNA in a sequence-independent manner and with a >30-fold fluorescence enhancement. It is useful for detecting nucleic acids in solution, or for selectively staining dead cells with damaged plasma membranes. Biotium offers Ethidium Homodimer I in much higher purity than other manufacturers. The dye has an excitation/emission of 528/617 nm when bound to DNA. Properties: Red solid soluble in water or DMSO; Store dessicated at 4°C, protect from light; Ex/Em = 528/617 nm (with DNA); C46H50Cl4N8; MW:857; [61926-22-5].