Learn More
Medchemexpress LLC EthD-III | 95.0% | 978.36 | 250 UG
SDP
Catalog No. 5000430476
Encompass_Preferred
EthD-III is a nucleic acid probe and a red fluorescent stain used to detect dead cells. It enters cells with damaged membranes and binds to nucleic acids, leading to bright red fluorescence in dead cells (Ex/Em=530/645 nm).
  • Nucleic acid probe
  • Red fluorescent stain
  • Detects dead cells
  • Enters cells with damaged membranes
  • Binds to nucleic acids
  • Results in bright red fluorescence in dead cells
Catalog No. 50-004-30476 Supplier Medchemexpress LLC Supplier No. 250UGHYD1723
EthD-III is a nucleic acid probe and a red fluorescent stain used to detect dead cells. It enters cells with damaged membranes and binds to nucleic acids, leading to bright red fluorescence in dead cells (Ex/Em=530/645 nm).
  • Nucleic acid probe
  • Red fluorescent stain
  • Detects dead cells
  • Enters cells with damaged membranes
  • Binds to nucleic acids
  • Results in bright red fluorescence in dead cells