Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethenamine, N,N-dimethyl-2-nitro- | 1190-92-7 | 99.9% | 116.12 | 10 G
SDP

Catalog No. 5000400703
Encompass_Preferred

1-(Dimethylamino)-2-nitroethylene is a drug intermediate utilized in the synthesis of various active compounds. It is intended strictly for research purposes and is not for sale to patients.

  • Used as a drug intermediate for synthesizing active compounds.
  • Appearance: Brown to dark brown solid.
  • Storage: Powder is stable for 3 years at -20°C or 2 years at 4°C.
  • Storage: In solvent, it is stable for 6 months at -80°C or 1 month at -20°C.
  • Solubility: ≥ 200 mg/mL (1722.36 mM) in DMSO.
  • Documentation available: Data Sheet, Certificate of Analysis (COA), Safety Data Sheet (SDS), HNMR, GC.

Catalog No. 50-004-00703 Supplier Medchemexpress LLC Supplier No. HY2039510G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-(Dimethylamino)-2-nitroethylene is a drug intermediate utilized in the synthesis of various active compounds. It is intended strictly for research purposes and is not for sale to patients.

  • Used as a drug intermediate for synthesizing active compounds.
  • Appearance: Brown to dark brown solid.
  • Storage: Powder is stable for 3 years at -20°C or 2 years at 4°C.
  • Storage: In solvent, it is stable for 6 months at -80°C or 1 month at -20°C.
  • Solubility: ≥ 200 mg/mL (1722.36 mM) in DMSO.
  • Documentation available: Data Sheet, Certificate of Analysis (COA), Safety Data Sheet (SDS), HNMR, GC.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.