Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethene, 1,1-bis(methylthio)-2-nitro- | 13623-94-4 | MFCD00010443 | 99.7% | 165.23 g/mol | C4H7NO2S2 | 1 G
SDP

Catalog No. 5000301179
Encompass_Preferred

1,1-Bis(methylthio)-2-nitroethylene (CAS 13623-94-4) is an organic biochemical reagent used in life-science research. It is supplied as a solid with high purity and is commonly used as a building block or reagent in organic synthesis and biochemical studies.

  • High purity (99.7%).
  • Molecular formula C4H7NO2S2.
  • Molecular weight 165.23 g/mol.
  • Solid reagent for organic synthesis and biochemical research.
  • Available in lab-scale pack sizes starting at 1 g.

Catalog No. 50-003-01179 Supplier Medchemexpress LLC Supplier No. HYW0171711G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,1-Bis(methylthio)-2-nitroethylene (CAS 13623-94-4) is an organic biochemical reagent used in life-science research. It is supplied as a solid with high purity and is commonly used as a building block or reagent in organic synthesis and biochemical studies.

  • High purity (99.7%).
  • Molecular formula C4H7NO2S2.
  • Molecular weight 165.23 g/mol.
  • Solid reagent for organic synthesis and biochemical research.
  • Available in lab-scale pack sizes starting at 1 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.