Learn More
Medchemexpress LLC Ethephon-d4 | 1020719-29-2 | >98.0% | 148.52 g/mol | C2H2D4ClO3P | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW131845S1MG
Encompass_Preferred
Ethephon-d4 is the deuterium-labeled analogue of ethephon supplied as an analytical reference standard for research use. It is intended for use as an internal standard in mass spectrometry and quantitative assays to improve accuracy, sensitivity, and traceability when measuring ethephon in complex matrices. Supplied in small quantities suitable for analytical calibration and method development.
  • Deuterium-labeled internal standard for mass spectrometry.
  • 99 atom % D isotopic enrichment for clear mass separation.
  • Chemical purity >98% suitable for analytical reference use.
  • Compact quantities for trace-level calibration and method development.
  • Stable under recommended storage for consistent performance.
Catalog No. 50-002-83289