Learn More
Medchemexpress LLC Ethephon-d4 | 1020719-29-2 | >98.0% | 148.52 g/mol | C2H2D4ClO3P | 5 MG
SDP
Catalog No. 5000282847
Encompass_Preferred
Ethephon-d4 is the deuterium-labeled analogue of ethephon supplied as a stable-isotope reference material for tracer and quantitative analytical applications. It is provided as a white to off-white solid suitable for use as an internal standard in laboratory workflows, with storage and handling recommendations for low-temperature, sealed conditions.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS methods.
  • White to off-white solid, easy to handle.
  • High isotopic enrichment and chemical purity (>98.0%).
  • Supplied in pre-weighed milligram quantities for analytical use.
Catalog No. 50-002-82847 Supplier Medchemexpress LLC Supplier No. HYW131845S5MG
Ethephon-d4 is the deuterium-labeled analogue of ethephon supplied as a stable-isotope reference material for tracer and quantitative analytical applications. It is provided as a white to off-white solid suitable for use as an internal standard in laboratory workflows, with storage and handling recommendations for low-temperature, sealed conditions.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS methods.
  • White to off-white solid, easy to handle.
  • High isotopic enrichment and chemical purity (>98.0%).
  • Supplied in pre-weighed milligram quantities for analytical use.