Learn More
BINDER™ Ethernet Interface
SDP
Catalog No. 08111689 Shop All BINDER Inc. Products
Change view
Click to view available options
Material:
Plastic
Catalog No. Material
08-111-689 Plastic
1 options
Catalog No. 08-111-689 Supplier BINDER™ Supplier No. 80120990
Add to Cart
Add to Cart
For Use With: BINDER units BF 56, BF 115, BF 260
  • Instead of the RS 422 interface
  • For Multi Management Software APT-COM™
Specifications
Description Ethernet interface - for Multi Management Software APT-COM™
For Use With (Equipment) BINDER Chambers BF 56, BF 115, BF 260
Material Plastic
Type Ethernet Interface