Learn More
BINDER™ Ethernet Interface Converter Connection Cable Set
SDP
Catalog No. 08111772 Shop All BINDER Inc. Products
Change view
Click to view available options
Electrical Requirements:
120 V, 60 Hz
230 V, 50/60 Hz
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Electrical Requirements
08-111-772 230 V, 50/60 Hz
08-111-773 120 V, 60 Hz
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 08-111-772 Supplier BINDER™ Supplier No. 80120380
Only null left
Add to Cart
Add to Cart
Interface converter - RS 422 cable set and RS 422 / Ethernet interface converter for connection to 10-way plug distributor or for connection to a unit.
  • Set of connection cables and interface connector RS 422 or RS 232
  • For either 10 interfaces or one unit
Specifications
Description Interface converter - RS 422 cable set and RS 422 / Ethernet interface converter for connection to 10-way plug distributor or for connection to a unit
For Use With (Equipment) Multi Management Software APT-COM™
Material Plastic
Type Cable Set
Electrical Requirements 230 V, 50/60 Hz