Learn More
Electron Microscopy Sciences Earth-Friendly Sample Spoon, 1 tsp
SDP
Catalog No. 501927066
Encompass_Preferred
Perfect for retrieving small, specific volume samples of powders or liquids. Wide handles let you grip firmly and provide just enough stiffness to easily sample packed powders and other firm materials. Their unique flat bottom shape allows convenient dual use as weighing boats to save time and assure accuracy. Nonsterile and translucent in color.
Capacity: 1 tsp
Catalog No. 50-192-7066 Supplier Electron Microscopy Sciences Supplier No. 72133
Perfect for retrieving small, specific volume samples of powders or liquids. Wide handles let you grip firmly and provide just enough stiffness to easily sample packed powders and other firm materials. Their unique flat bottom shape allows convenient dual use as weighing boats to save time and assure accuracy. Nonsterile and translucent in color.
Capacity: 1 tsp