Learn More
Med Vet International Ethicon Vicryl Suture, Size 4-0, P-3, 18in, 12/Box
Catalog No. NC2203474
Encompass_Preferred
The Ethicon J494G suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 18in. in length, has a size of 4-0, they are undyed and typically have an absorption rate of 56-70 days. This suture comes with a P-3 Precision Point reverse cutting needle and has 12 sutures in a box.
This item is not returnable. View return policy
Catalog No. NC2203474 Supplier Med Vet International Supplier No. J494G
The Ethicon J494G suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 18in. in length, has a size of 4-0, they are undyed and typically have an absorption rate of 56-70 days. This suture comes with a P-3 Precision Point reverse cutting needle and has 12 sutures in a box.