Learn More
Med Vet International Ethicon Prolene Polypropylene Suture, Size 0, CT-1, 30in, 36/Box
Supplier:  Med Vet International 8424HRL
Encompass_Preferred
The Ethicon 8424H suture is a high-quality non-absorbable Polypropylene surgical suture. It measures 30in. in length, has a size of 0, and are dyed black to provide greater visibility while suturing. This suture comes with a CT-1 taper point needle and has 30 sutures in a box.
Catalog No. NC1841205
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.