Learn More
IBI SCIENTIFIC 3P Ethidium Bromide Solution | 1239-45-8 | MFCD00011724 | 10 mL
Supplier:  IBI SCIENTIFIC 3P IB40075
Encompass_Preferred
Ethidium Bromide Solution - 10ml IBI ethidium bromide can be used to detect both single stranded and double stranded nucleic acids (both DNA and RNA). However, ethidium bromide does not intercalate into single stranded DNA very well;
Catalog No. NC0753494