Learn More
Apexbio Technology LLC Ethidium bromide 1239-45-8 250mg
SDP
Catalog No. 5000088469
Encompass_Preferred
Ethidium bromide (CAS 1239-45-8) is a small-molecule fluorescent dye widely utilized in molecular biology for nucleic acid visualization It intercalates between the base pairs of double-stranded DNA or RNA resulting in enhanced fluorescence upon binding With an excitation spectrum between 300 360 nm and an emission maximum at 590 nm ethidium bromide is commonly used to stain nucleic acids during agarose gel electrophoresis This property facilitates the detection and analysis of DNA and RNA in applications such as gene expression studies and nucleic acid quantification supporting a broad range of experimental workflows in genomics and molecular diagnostics
Catalog No. 50-000-88469 Supplier Apexbio Technology LLC Supplier No. C6568250
Ethidium bromide (CAS 1239-45-8) is a small-molecule fluorescent dye widely utilized in molecular biology for nucleic acid visualization It intercalates between the base pairs of double-stranded DNA or RNA resulting in enhanced fluorescence upon binding With an excitation spectrum between 300 360 nm and an emission maximum at 590 nm ethidium bromide is commonly used to stain nucleic acids during agarose gel electrophoresis This property facilitates the detection and analysis of DNA and RNA in applications such as gene expression studies and nucleic acid quantification supporting a broad range of experimental workflows in genomics and molecular diagnostics