Learn More
Medchemexpress LLC Ethidium bromide | 1239-45-8 | MFCD00011724 | 99.4% | 100 MG
SDP
Catalog No. 5000410988
Encompass_Preferred
EthD bromide is an intercalating agent commonly used as a fluorescent tag (nucleic acid stain) in molecular biology laboratories for techniques such as agarose gel electrophoresis. It is for research use only and not sold to patients. It binds to DNA, allowing visualization under ultraviolet (UV) light.
  • Serves as a fluorescent tag (nucleic acid stain)
  • Used in molecular biology laboratories for techniques such as agarose gel electrophoresis
  • Binds to DNA
  • Allows visualization under ultraviolet (UV) light
Catalog No. 50-004-10988 Supplier Medchemexpress LLC Supplier No. HYD0021100MG
EthD bromide is an intercalating agent commonly used as a fluorescent tag (nucleic acid stain) in molecular biology laboratories for techniques such as agarose gel electrophoresis. It is for research use only and not sold to patients. It binds to DNA, allowing visualization under ultraviolet (UV) light.
  • Serves as a fluorescent tag (nucleic acid stain)
  • Used in molecular biology laboratories for techniques such as agarose gel electrophoresis
  • Binds to DNA
  • Allows visualization under ultraviolet (UV) light