Learn More
Bioworld Ethidium Bromide, 25 g
Catalog No. 50255962
Encompass_Preferred
Biotechnology GradeA fluorescent electrophoretic stain for nucleic acidsAlso used as a nucleic acid intercalating agent and frameshift mutagenAmmonium bromide Concentration: <0.1%Regulated SubstanceFluoresence: λex 480 nm, λem 620 nm in waterHaz Mat fee appliesAppearance: SolidHS Code : 29339990Melting Point: 260-262 °C (dec.)(lit.)RTECS Number: SF7950000
Catalog No. 50-255-962 Supplier Bioworld Supplier No. 405400052
Biotechnology GradeA fluorescent electrophoretic stain for nucleic acidsAlso used as a nucleic acid intercalating agent and frameshift mutagenAmmonium bromide Concentration: <0.1%Regulated SubstanceFluoresence: λex 480 nm, λem 620 nm in waterHaz Mat fee appliesAppearance: SolidHS Code : 29339990Melting Point: 260-262 °C (dec.)(lit.)RTECS Number: SF7950000