Learn More
Selleck Chemical LLC Ethidium bromide S3689-25mg
SDP
Catalog No. 503110522
Encompass_Preferred
Ethidium Bromide (Homidium bromide EtBr EB) is an intercalating agent which resembles a DNA base pair and commonly used as a fluorescent tag (nucleic acid stain) in molecular biology laboratories for techniques such as agarose gel electrophoresis
Catalog No. 50-311-0522 Supplier Selleck Chemical LLC Supplier No. S368925MG
Ethidium Bromide (Homidium bromide EtBr EB) is an intercalating agent which resembles a DNA base pair and commonly used as a fluorescent tag (nucleic acid stain) in molecular biology laboratories for techniques such as agarose gel electrophoresis