Learn More
Research Products International Corp RPI Ethidium Bromide, Powder, 25 Grams
SDP
Supplier:  Research Products International Corp E7180025.0
Encompass_Preferred
Ethidium Bromide is commonly used as a nucleic acid stain for gel electrophoresis due to its fluorescence when exposed to UV light. Ethidium Bromide intercalates double-stranded DNA and RNA and also acts as inhibitor of RNA and DNA polymerase. Can be used to stain gel after electrophoresis or incorporated into the gel and running buffer.
Catalog No. 50-197-7193