Learn More
Bioworld Ethidium Bromide Dropper Bottle, 5 mL
Catalog No. 50255960
Encompass_Preferred
ml> false false false ml> Ethidium bromide, dropper bottles A fluorescent electrophoretic stain for nucleic acids. It is used as a nucleic acid intercalating agent and frameshift mutagen. Ethidium Bromide solution in dropper form makes dispensing safer and convenient to use. Each drop contains about 25µg of Ethidium Bromide and is perfectly suited for a 50ml agarose gel. Simply add one drop of the Ethidium Bromide solution per 50ml gel solution (before gelation) and cast your gel. Technical Specifications:Biotechnology Grade Concentration: 0.636µg/µL stock final concentration: 5ml> false false false ml> µg/mL Appearance: LiquidHS Code : 29339990Melting Point: 260-262 °C (dec.)(lit.)RTECS Number: SF7950000
Catalog No. 50-255-960 Supplier Bioworld Supplier No. 405400041
ml> false false false ml> Ethidium bromide, dropper bottles A fluorescent electrophoretic stain for nucleic acids. It is used as a nucleic acid intercalating agent and frameshift mutagen. Ethidium Bromide solution in dropper form makes dispensing safer and convenient to use. Each drop contains about 25µg of Ethidium Bromide and is perfectly suited for a 50ml agarose gel. Simply add one drop of the Ethidium Bromide solution per 50ml gel solution (before gelation) and cast your gel. Technical Specifications:Biotechnology Grade Concentration: 0.636µg/µL stock final concentration: 5ml> false false false ml> µg/mL Appearance: LiquidHS Code : 29339990Melting Point: 260-262 °C (dec.)(lit.)RTECS Number: SF7950000