Learn More
Genesee Scientific Corporation ETHIDIUM BROMIDE DROPPER BOTTL
SDP
Catalog No. NC0100518
Encompass
Ethidium Bromide, 0.625mg/ml 1 Bottle/Unit Convenient and safe alternative 10ml, 0.625mg/ml Simply add one drop for every 50ml of solution for a final concentration of 0.5ug/ml Shelf life: When stored properly, indefinite. As a precaution, 3 years from date of purchase.
Catalog No. NC0100518 Supplier Genesee Scientific Corporation Supplier No. 20276
Ethidium Bromide, 0.625mg/ml 1 Bottle/Unit Convenient and safe alternative 10ml, 0.625mg/ml Simply add one drop for every 50ml of solution for a final concentration of 0.5ug/ml Shelf life: When stored properly, indefinite. As a precaution, 3 years from date of purchase.