Learn More
Research Products International Corp Ethidium Bromide, Powder, 1 Gram
SDP
Catalog No. 501978405
Encompass_Preferred
Ethidium Bromide is commonly used as a nucleic acid stain for gel electrophoresis due to its fluorescence when exposed to UV light. Ethidium Bromide intercalates double-stranded DNA and RNA and also acts as inhibitor of RNA and DNA polymerase. Can be used to stain gel after electrophoresis or incorporated into the gel and running buffer.
Catalog No. 50-197-8405 Supplier Research Products International Corp Supplier No. E718001.0
Ethidium Bromide is commonly used as a nucleic acid stain for gel electrophoresis due to its fluorescence when exposed to UV light. Ethidium Bromide intercalates double-stranded DNA and RNA and also acts as inhibitor of RNA and DNA polymerase. Can be used to stain gel after electrophoresis or incorporated into the gel and running buffer.