Learn More
Alkali Scientific Ethidium Bromide, Powder, 5 G
Supplier: Alkali Scientific HID001CP
Ethidium Bromide (EtBr), 5g, is a highly effective nucleic acid stain used in molecular biology for DNA visualization under UV light. As an intercalating agent, Ethidium Bromide binds to DNA and emits a fluorescent signal when exposed to ultraviolet light, allowing researchers to easily detect and analyze DNA bands in electrophoresis gels. This powder form is convenient for preparing various concentrations of staining solutions depending on experimental needs. Ethidium Bromide is widely used in genetic research, PCR analysis, and gel electrophoresis for visualizing DNA, making it a fundamental reagent in molecular and cellular biology laboratories.