Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Ethidium Bromide Solution (10mg/mL)

Catalog No. MP1ETBC1002 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mL
6 x 10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP1ETBC1002 6 x 10 mL
MP1ETBC1001 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP1ETBC1002 Supplier MP Biomedicals™ Supplier No. 11ETBC1002
Add to Cart
Add to Cart

Ethidium Bromide Solution (10mg/mL), Molecular biology grade

  • Ethidium bromide (EtBr) is useful for all applications requiring nucleic acid staining including CsCl plasmid DNA isolation and is the most commonly used nucleic acid stain for PAGE or agarose gel electrophoresis.
  • Ethidium bromide has been used in a number of fluorimetric assays for nucleic acids.
  • A 10 mg/mL Ready-To-Use solution of EtBr in specially filtered, deionized water.
  • Ethidium bromide (EtBr) is useful for all applications requiring nucleic acid staining including CsCl plasmid DNA isolation and is the most commonly used nucleic acid stain for PAGE or agarose gel electrophoresis.
  • It has been shown to bind to single-stranded DNA (although not as strongly) and triple-stranded DNA.
  • MP's EtBr confers a deep redstain to DNA by acting as an intercalating agent between the starch bases.
  • Because of its ability to bind to DNA, EtBr is an inhibitor of DNA polymerase.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1239-45-8
Detection 15-30 min with 0.5mg/mL solution
Color Dark Red/Purple
Concentration 10 mg/mL
Form Liquid
For Use With (Application) Nucleic Acid Electrophoresis; Blotting & Detection; Nucleic Acidgel Electrophoresis & Blotting
Molecular Weight (g/mol) 394.3
Quantity 6 x 10 mL
Storage Requirements Store at +4°C, protect from light.
Product Type Fluorescent DNA/RNA Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.