Learn More
AdipoGen Ethidium homodimer, 61926-22-5, MFCD18433392, 1 mg
SDP
Catalog No. 501147082
Encompass_Preferred
MF: C46H50Cl4N8, MW: 856.75, ~90%, Appearance: Dark red solid. Staining dye for ssDNA, dsDNA, RNA, oligonucleotides and triplex DNA. It does not cross intact cell membranes and can be used to test cell viability. Reagent for the fluorimetric detection of nucleic acids.
Catalog No. 50-114-7082 Supplier AdipoGen Supplier No. CDXE0012M001
MF: C46H50Cl4N8, MW: 856.75, ~90%, Appearance: Dark red solid. Staining dye for ssDNA, dsDNA, RNA, oligonucleotides and triplex DNA. It does not cross intact cell membranes and can be used to test cell viability. Reagent for the fluorimetric detection of nucleic acids.