Learn More
AdipoGen Ethidium homodimer
SDP
Catalog No. 502560997
Encompass_Preferred
Chemical. CAS 61926-22-5. Formula C46H50Cl4N8. MW 856.75. Synthetic. Staining dye for ssDNA, dsDNA, RNA, oligonucleotides and triplex DNA. It does not cross intact cell membranes and can be used to test cell viability. Reagent for the fluorimetric detection of nucleic acids.
Catalog No. 50-256-0997 Supplier AdipoGen Supplier No. CDXE0012M010
Chemical. CAS 61926-22-5. Formula C46H50Cl4N8. MW 856.75. Synthetic. Staining dye for ssDNA, dsDNA, RNA, oligonucleotides and triplex DNA. It does not cross intact cell membranes and can be used to test cell viability. Reagent for the fluorimetric detection of nucleic acids.