Learn More
Abcam Ethidium homodimer, Fluorimetric detection reagent, 1MG
Catalog No. 502705289
Encompass_Preferred
MW 856.75 g/mol, Purity =90%. High-affinity nucleic acid stain for ssDNA, dsDNA, RNA, oligonucleotides and triplex DNA. Weakly fluorescent until bound to DNA. It does not cross intact cell membranes and can be used to test cell viability.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5289 Supplier Abcam Supplier No. AB1453231MG
MW 856.75 g/mol, Purity =90%. High-affinity nucleic acid stain for ssDNA, dsDNA, RNA, oligonucleotides and triplex DNA. Weakly fluorescent until bound to DNA. It does not cross intact cell membranes and can be used to test cell viability.
The product is subject to the following: Abcam Restricted Use Statement