Learn More
Apexbio Technology LLC Ethinyl Estradiol 57-63-6 100mg
SDP
Supplier:  Apexbio Technology LLC A8428100
Encompass_Preferred
Ethinyl Estradiol (57-63-6) is a small-molecule estrogen receptor agonist targeting intracellular estrogen receptors It is designed to activate these receptors thereby regulating transcription of estrogen-responsive genes and influencing reproductive biology and cell proliferation pathways Ethinyl Estradiol exerts its biological activity primarily through direct binding to estrogen receptors leading to the modulation of endocrine-regulated mechanisms Based on these pharmacological properties Ethinyl Estradiol holds research potential in studies of receptor-mediated responses endocrine regulation mechanisms synthetic hormone interactions drug interactions contraceptive mechanisms and hormone-dependent physiological processes
Catalog No. 50-000-96237