Learn More
Selleck Chemical LLC Ethinyl Estradiol S1625-50mg
SDP
Catalog No. 503111304
Encompass_Preferred
Ethinyl Estradiol is an orally bio-active estrogen used in almost all modern formulations of combined oral contraceptive pills
Catalog No. 50-311-1304 Supplier Selleck Chemical LLC Supplier No. S162550MG
Ethinyl Estradiol is an orally bio-active estrogen used in almost all modern formulations of combined oral contraceptive pills