Learn More
Selleck Chemical LLC Ethinyl Estradiol S1625-1g
SDP
Catalog No. 503111307
Encompass_Preferred
Ethinyl Estradiol is an orally bio-active estrogen used in almost all modern formulations of combined oral contraceptive pills
Catalog No. 50-311-1307 Supplier Selleck Chemical LLC Supplier No. S16251G
Add to Cart
Add to Cart
Ethinyl Estradiol is an orally bio-active estrogen used in almost all modern formulations of combined oral contraceptive pills