Learn More
Apexbio Technology LLC Ethionamide 536-33-4 1g
SDP
Catalog No. 5000093861
Encompass_Preferred
Ethionamide (CAS 536-33-4) is a small-molecule inhibitor targeting enoyl-acyl carrier protein reductase (InhA) It is designed to inhibit mycolic acid biosynthesis thereby disrupting bacterial cell-wall integrity Ethionamide exerts its biological activity primarily through inhibiting the function of InhA In in vitro studies Ethionamide demonstrates antimycobacterial activity against Mycobacterium tuberculosis with reported MIC values typically ranging from approximately 0 3 to 2 5 g/mL Based on these pharmacological properties Ethionamide holds research potential in studies of multidrug- or extensively drug-resistant tuberculosis
Catalog No. 50-000-93861 Supplier Apexbio Technology LLC Supplier No. A84291000
Ethionamide (CAS 536-33-4) is a small-molecule inhibitor targeting enoyl-acyl carrier protein reductase (InhA) It is designed to inhibit mycolic acid biosynthesis thereby disrupting bacterial cell-wall integrity Ethionamide exerts its biological activity primarily through inhibiting the function of InhA In in vitro studies Ethionamide demonstrates antimycobacterial activity against Mycobacterium tuberculosis with reported MIC values typically ranging from approximately 0 3 to 2 5 g/mL Based on these pharmacological properties Ethionamide holds research potential in studies of multidrug- or extensively drug-resistant tuberculosis