Learn More
Cayman Chemical Ethionamide-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 2748267-66-3, 1 mg
SDP
Catalog No. 502948900
Encompass_Preferred
An internal standard for the quantification of ethionamide by GC- or LC-MS
Catalog No. 50-294-8900 Supplier Cayman Chemical Supplier No. 336361 MG
An internal standard for the quantification of ethionamide by GC- or LC-MS