Learn More
Cayman Chemical EthIonamIde-d3 500ug
SDP
Catalog No. 502947894
Encompass_Preferred
An internal standard for the quantification of ethionamide by GC- or LC-MS
Catalog No. 50-294-7894 Supplier Cayman Chemical Supplier No. 33636500 UG
An internal standard for the quantification of ethionamide by GC- or LC-MS