Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethisterone 98.0+%, TCI America™
SDP

Catalog No. E00401G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E00401G 1 g
E00405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E00401G Supplier TCI America Supplier No. E00401G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 434-03-7
Molecular Formula C21H28O2
Molecular Weight (g/mol) 312.453
MDL Number MFCD00003656
InChI Key CHNXZKVNWQUJIB-SJFWLOONSA-N
Synonym pregn-4-en-20-yn-3-one, 17-hydroxy-, 17a, ksc587g1r, 17-hydroxypregna-4-ene-20-yne-3-one
PubChem CID 12016247
IUPAC Name (8R,9S,10R,13S,14S,17S)-17-ethynyl-17-hydroxy-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
SMILES CC12CCC(=O)C=C1CCC3C2CCC4(C3CCC4(C#C)O)C

Specifications

Melting Point 272°C
Quantity 1 g
Formula Weight 312.45
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethisterone
RTECSNumber TU5570250
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.