Learn More
Cayman Chemical Ethionamide, Purity: ≥98%, CAS: 536-33-4, 10 g
SDP
Catalog No. 502995049
Encompass_Preferred
An antimycobacterial compound active against M. tuberculosis (MICs = 0.3-1.25 µg/ml); activated via oxidation by flavin monooxygenase; inhibits InhA to block mycolic acid biosynthesis
Catalog No. 50-299-5049 Supplier Cayman Chemical Supplier No. 2371410 G
An antimycobacterial compound active against M. tuberculosis (MICs = 0.3-1.25 µg/ml); activated via oxidation by flavin monooxygenase; inhibits InhA to block mycolic acid biosynthesis