Learn More
Medchemexpress LLC Ethopropazine hydrochloride | 1094-08-2 | MFCD00012653 | 99.3% | 348.93 | C19H25ClN2S | 1 ML
SDP
Catalog No. 5000241781
Encompass_Preferred
Ethopropazine hydrochloride is a research-grade anticholinergic compound used in neuroscience and pharmacology studies. It functions primarily as a butyrylcholinesterase inhibitor, with weaker activity against acetylcholinesterase, and exhibits non-selective muscarinic and NMDA receptor antagonism. The compound is used for investigations of cholinergic signaling and in Parkinson's disease models, and is supplied in formats suitable for in vitro dosing and analytical work.
  • Potent butyrylcholinesterase (BChE) inhibitor.
  • Weak acetylcholinesterase (AChE) activity for selectivity studies.
  • Non-selective muscarinic and NMDA receptor antagonism.
  • Suitable for Parkinson's disease and neuropharmacology research.
  • Available as solid or pre-made 10 mM solution for easy dosing.
  • Documented stability and storage guidelines for reproducible results.
Catalog No. 50-002-41781 Supplier Medchemexpress LLC Supplier No. HY10792210MM 1ML
Ethopropazine hydrochloride is a research-grade anticholinergic compound used in neuroscience and pharmacology studies. It functions primarily as a butyrylcholinesterase inhibitor, with weaker activity against acetylcholinesterase, and exhibits non-selective muscarinic and NMDA receptor antagonism. The compound is used for investigations of cholinergic signaling and in Parkinson's disease models, and is supplied in formats suitable for in vitro dosing and analytical work.
  • Potent butyrylcholinesterase (BChE) inhibitor.
  • Weak acetylcholinesterase (AChE) activity for selectivity studies.
  • Non-selective muscarinic and NMDA receptor antagonism.
  • Suitable for Parkinson's disease and neuropharmacology research.
  • Available as solid or pre-made 10 mM solution for easy dosing.
  • Documented stability and storage guidelines for reproducible results.