Learn More
Medchemexpress LLC Ethopropazine hydrochloride | 1094-08-2 | MFCD00012653 | 99.3% | 348.93 g·mol⁻¹ | C19H25ClN2S | 10 MG
SDP
Catalog No. 5000241720
Encompass_Preferred
Ethopropazine hydrochloride is a research chemical with anticholinergic properties. It acts as a butyrylcholinesterase (BChE) inhibitor, is a poor acetylcholinesterase (AChE) inhibitor, and functions as a non-selective muscarinic acetylcholine receptor (mAChR) and NMDA antagonist. It is used in pharmacology and neuroscience research, including studies related to Parkinson's disease.
  • High reported purity suitable for research applications.
  • Characterized pharmacological profile for cholinergic and glutamatergic studies.
  • Available in small-mass vials for dosing and analytical work.
  • Provided as the hydrochloride salt for improved solubility in aqueous media.
  • Useful in Parkinson's disease and neurodegeneration research models.
Catalog No. 50-002-41720 Supplier Medchemexpress LLC Supplier No. HY10792210MG
Ethopropazine hydrochloride is a research chemical with anticholinergic properties. It acts as a butyrylcholinesterase (BChE) inhibitor, is a poor acetylcholinesterase (AChE) inhibitor, and functions as a non-selective muscarinic acetylcholine receptor (mAChR) and NMDA antagonist. It is used in pharmacology and neuroscience research, including studies related to Parkinson's disease.
  • High reported purity suitable for research applications.
  • Characterized pharmacological profile for cholinergic and glutamatergic studies.
  • Available in small-mass vials for dosing and analytical work.
  • Provided as the hydrochloride salt for improved solubility in aqueous media.
  • Useful in Parkinson's disease and neurodegeneration research models.