Learn More
Medchemexpress LLC Ethopropazine hydrochloride | 1094-08-2 | MFCD00012653 | 99.3% | 348.93 g/mol | C19H25ClN2S | 5 MG
SDP
Catalog No. 5000242914
Encompass_Preferred
Ethopropazine hydrochloride is the monohydrochloride salt of ethopropazine, a phenothiazine-derived research compound with anticholinergic activity. It is commonly used in neuroscience research as a butyrylcholinesterase inhibitor and as a non-selective muscarinic and NMDA antagonist. The compound is provided as a white to off-white solid and as DMSO solutions for laboratory use.
  • Potent BChE inhibitor for cholinesterase studies.
  • Non-selective mAChR and NMDA antagonist for receptor pharmacology.
  • High purity suitable for analytical and research applications.
  • Available as solid powder and ready-to-use DMSO solution for flexibility.
  • Stable when stored sealed, away from moisture and light under recommended conditions.
Catalog No. 50-002-42914 Supplier Medchemexpress LLC Supplier No. HY1079225MG
Ethopropazine hydrochloride is the monohydrochloride salt of ethopropazine, a phenothiazine-derived research compound with anticholinergic activity. It is commonly used in neuroscience research as a butyrylcholinesterase inhibitor and as a non-selective muscarinic and NMDA antagonist. The compound is provided as a white to off-white solid and as DMSO solutions for laboratory use.
  • Potent BChE inhibitor for cholinesterase studies.
  • Non-selective mAChR and NMDA antagonist for receptor pharmacology.
  • High purity suitable for analytical and research applications.
  • Available as solid powder and ready-to-use DMSO solution for flexibility.
  • Stable when stored sealed, away from moisture and light under recommended conditions.