Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethopropazine hydrochloride | 1094-08-2 | MFCD00012653 | 99.3% | 348.93 g/mol | C19H25ClN2S | 50 MG
SDP

Catalog No. 5000242657
Encompass_Preferred

Ethopropazine hydrochloride is the hydrochloride salt of a phenothiazine-class compound used in research as a butyrylcholinesterase (BChE) inhibitor and a non-selective muscarinic and NMDA antagonist. It is applied in neuroscience studies, including Parkinson's disease and analgesia models, for its anticholinergic and neuropharmacological effects.

  • High purity: 99.3%.
  • Molecular weight: 348.93 g/mol.
  • Chemical formula: C19H25ClN2S.
  • CAS number: 1094-08-2.
  • Available as a 50 mg research pack.
  • Useful as a BChE inhibitor in neuropharmacology studies.

Catalog No. 50-002-42657 Supplier Medchemexpress LLC Supplier No. HY10792250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethopropazine hydrochloride is the hydrochloride salt of a phenothiazine-class compound used in research as a butyrylcholinesterase (BChE) inhibitor and a non-selective muscarinic and NMDA antagonist. It is applied in neuroscience studies, including Parkinson's disease and analgesia models, for its anticholinergic and neuropharmacological effects.

  • High purity: 99.3%.
  • Molecular weight: 348.93 g/mol.
  • Chemical formula: C19H25ClN2S.
  • CAS number: 1094-08-2.
  • Available as a 50 mg research pack.
  • Useful as a BChE inhibitor in neuropharmacology studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.