Learn More
Cayman Chemical Ethopropazine (hydrochloride), Purity: ≥98%, CAS: 1094-08-2, 50 mg
SDP
Catalog No. 502939530
Encompass_Preferred
A BChE inhibitor (IC50 = 15.14 µM in human erythrocyte lysates); selective for BChE over AChE at 500 µM; binds to rat forebrain and hindbrain membrane preparations (Kis = 3.1 and 7.2 nM, respectively), which are endogenously enriched in M1 and M2 muscarinic acetylcholine receptors, respectively; decreases the intensity and increases the latency of haloperidol-induced catalepsy, a model of extrapyramidal syndrome, in rats when used in combination with ZM 241395 at 0.232 mg/kg; reduces thermal hyperalgesia in a rat model of neuropathic pain induced by sciatic nerve ligation at 20 and 30 mg/kg
Catalog No. 50-293-9530 Supplier Cayman Chemical Supplier No. 2849350 MG
A BChE inhibitor (IC50 = 15.14 µM in human erythrocyte lysates); selective for BChE over AChE at 500 µM; binds to rat forebrain and hindbrain membrane preparations (Kis = 3.1 and 7.2 nM, respectively), which are endogenously enriched in M1 and M2 muscarinic acetylcholine receptors, respectively; decreases the intensity and increases the latency of haloperidol-induced catalepsy, a model of extrapyramidal syndrome, in rats when used in combination with ZM 241395 at 0.232 mg/kg; reduces thermal hyperalgesia in a rat model of neuropathic pain induced by sciatic nerve ligation at 20 and 30 mg/kg