Learn More
Medchemexpress LLC Ethosuximide-d3 | 1189703-33-0 | ≥99.0% | 1 MG
SDP
Catalog No. 5000412775
Encompass_Preferred
Ethosuximide-d3 is the deuterium labeled Ethosuximide. It is a widely prescribed anti-epileptic agent that improves the phenotypes of multiple neurodegenerative disease models and blocks the low voltage activated T-type calcium channel. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled ethosuximide.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Improves phenotypes of multiple neurodegenerative disease models.
  • Blocks low voltage activated T-type calcium channel.
  • Deuterium substitution can affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-12775 Supplier Medchemexpress LLC Supplier No. HYB1378S1MG
Ethosuximide-d3 is the deuterium labeled Ethosuximide. It is a widely prescribed anti-epileptic agent that improves the phenotypes of multiple neurodegenerative disease models and blocks the low voltage activated T-type calcium channel. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled ethosuximide.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Improves phenotypes of multiple neurodegenerative disease models.
  • Blocks low voltage activated T-type calcium channel.
  • Deuterium substitution can affect pharmacokinetic and metabolic profiles of drugs.