Learn More
Cayman Chemical Ethosuximide-d5, Purity: ≥99% deuterated forms (d1-d5), CAS: 1989660-59-4, 5 mg
SDP
Catalog No. 502988192
Encompass_Preferred
An internal standard for the quantification of ethosuximide by GC- or LC-MS
Catalog No. 50-298-8192 Supplier Cayman Chemical Supplier No. 307595 MG
An internal standard for the quantification of ethosuximide by GC- or LC-MS