Learn More
Medchemexpress LLC Ethotoin | 86-35-1 | MFCD00072127 | 99.6% | 25 MG
SDP
Catalog No. 5000414079
Encompass_Preferred
Ethotoin is an orally active anticonvulsant agent used in epilepsy research. It is a hydantoin, similar to phenytoin. It is supplied as a solid, white to off-white in color, with a purity of 99.61%.
Catalog No. 50-004-14079 Supplier Medchemexpress LLC Supplier No. 25MGHYB1642
Ethotoin is an orally active anticonvulsant agent used in epilepsy research. It is a hydantoin, similar to phenytoin. It is supplied as a solid, white to off-white in color, with a purity of 99.61%.