Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethoxyacetic acid | 627-03-2 | MFCD00004310 | 98.0% | 104.10 | C4H8O3 | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHYW027555

Encompass_Preferred

Ethoxyacetic acid is an endogenous metabolite provided as a research-grade analytical standard. The compound is specified for high purity and is characterized by the molecular formula C4H8O3 and molecular weight 104.10 g/mol, making it suitable for analytical, toxicological, and metabolite-profiling studies.

  • High purity (98.0%) suitable for analytical applications.
  • Characterized molecular weight 104.10 g/mol and formula C4H8O3.
  • Available in multiple pack sizes, including 25 g, for flexible experimental needs.
  • Soluble in DMSO, facilitating preparation of concentrated stock solutions.
  • Supplied with safety data sheet and batch-specific documentation.

Catalog No. 50-003-03381


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.