Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethoxyacetic acid | 627-03-2 | MFCD00004310 | 98.0% | 104.11 g/mol | C4H8O3 | 10 G
SDP

Catalog No. 5000301944
Encompass_Preferred

Ethoxyacetic acid is an endogenous metabolite supplied as a high-purity analytical standard for research use. It is provided as a colorless to light-yellow liquid (C4H8O3) with characterized molecular weight and documented quality data, intended for biochemical, analytical, and formulation studies.

  • Purity 98.0% (NMR) for analytical reliability.
  • Molecular weight 104.11 g/mol; formula C4H8O3 for accurate calculations.
  • High solubility in DMSO (75 mg/mL) for easy preparation of stock solutions.
  • Available in multiple liquid package sizes suitable for lab-scale use.
  • Batch-specific certificate of analysis and SDS available for quality control.

Catalog No. 50-003-01944 Supplier Medchemexpress LLC Supplier No. HYW02755510G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethoxyacetic acid is an endogenous metabolite supplied as a high-purity analytical standard for research use. It is provided as a colorless to light-yellow liquid (C4H8O3) with characterized molecular weight and documented quality data, intended for biochemical, analytical, and formulation studies.

  • Purity 98.0% (NMR) for analytical reliability.
  • Molecular weight 104.11 g/mol; formula C4H8O3 for accurate calculations.
  • High solubility in DMSO (75 mg/mL) for easy preparation of stock solutions.
  • Available in multiple liquid package sizes suitable for lab-scale use.
  • Batch-specific certificate of analysis and SDS available for quality control.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.