Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethoxyacetylene, ca 50% w/w in hexanes

Catalog No. AAA1424506
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1424506 5 g
AAA1424503 1 g
AAA1424514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1424506 Supplier Thermo Scientific Chemicals Supplier No. A1424506
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 927-80-0 | C4H6O | 70.091 g/mol

Ethoxyacetylene is used in Arens-van Dorp synthesis to prepare propargyl alcohol via reaction with ketone. In organic synthesis, it undergoes [2+2] cycloaddition reactions with ketenes to give cyclobutenone derivatives. It is also used to prepare alpha, beta-unsaturated esters from ketones by following Meyer-Schuster rearrangement.Used with N-vinylamides and triflic anhydride in a direct, three-component synthesis of pyridines. Also used to prepare α, β-unsaturated esters from ketones via a Meyer-Schuster rearrangement.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethoxyacetylene is used in Arens-van Dorp synthesis to prepare propargyl alcohol via reaction with ketone. In organic synthesis, it undergoes [2+2] cycloaddition reactions with ketenes to give cyclobutenone derivatives. It is also used to prepare alpha, beta-unsaturated esters from ketones by following Meyer-Schuster rearrangement.Used with N-vinylamides and triflic anhydride in a direct, three-component synthesis of pyridines. Also used to prepare α, β-unsaturated esters from ketones via a Meyer-Schuster rearrangement.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 927-80-0
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.091
MDL Number MFCD00009247
InChI Key WMYNMYVRWWCRPS-UHFFFAOYSA-N
Synonym ethoxyacetylene, ethoxyethyne, ethyl ethynyl ether, ethyne, ethoxy, ether, ethyl ethynyl, ethoxy-ethyne, ethyne, ethoxy-9ci, 1-ethoxyacetylene, ethoxyacetylene w/w solution in hexanes, ethoxy acetylene
PubChem CID 61239
IUPAC Name ethynoxyethane
SMILES CCOC#C

Specifications

Density 0.718
Boiling Point 55°C to 56°C
Flash Point −28°C (−18°F)
Odor Recognizable
Refractive Index 1.38
Linear Formula CH3CH2OC≡CH
Quantity 5 g
UN Number UN1993
Beilstein 741882
Solubility Information Slightly miscible with water.
Formula Weight 70.09
Concentration or Composition (by Analyte or Components) ≈50% w/w in hexanes
Chemical Name or Material Ethoxyacetylene
Show More Show Less
Hazard Category H225-H304-H312+H332-H315-H319-H336-H351-H361f-H373
Hazard Statement GHS H Statement
H225-H304-H361-H373-H315
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 213-164-5
RTECSNumber KN9900000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.