Learn More
Medchemexpress LLC Ethoxybenzene | 103-73-1 | MFCD00009090 | 99.9% | 122.16 g/mol | C8H10O | 1kg
SDP
Catalog No. 5000266646
Encompass_Preferred
Ethoxybenzen can be used as an analyte to determine the properties of porous graphitic carbon (PGC) particles[1]
Catalog No. 50-002-66646 Supplier Medchemexpress LLC Supplier No. HYY12071KG
Ethoxybenzen can be used as an analyte to determine the properties of porous graphitic carbon (PGC) particles[1]