Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethoxydiphenylphosphine 93.0+%, TCI America™
SDP

Catalog No. D25585G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25585G 5 g
D255825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25585G Supplier TCI America Supplier No. D25585G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 719-80-2
Molecular Formula C14H15OP
Molecular Weight (g/mol) 230.25
MDL Number MFCD00009087
InChI Key JCRCPEDXAHDCAJ-UHFFFAOYSA-N
Synonym ethyl diphenylphosphinite, ethoxydiphenylphosphine, ethyldiphenylphosphinite, phosphinous acid, diphenyl-, ethyl ester, ethyl-diphenylphosphinit, pubchem6468, diphenyethoxy phosphine, ethoxydiphenyl phosphine, acmc-1bbad, ethoxy diphenyl phosphane
PubChem CID 69754
IUPAC Name ethyl diphenylphosphinite
SMILES CCOP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 230.25
Percent Purity ≥93.0% (T)
Physical Form Liquid
Chemical Name or Material Ethoxydiphenylphosphine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.