Learn More
Medchemexpress LLC Ethoxymethyl benzene | 539-30-0 | MFCD00039903 | 99.0% | 136.19 | C9H12O | 1 ML
SDP
Catalog No. 5000301817
Encompass_Preferred
(Ethoxymethyl)benzene is an endogenous metabolite used as an analytical reference and reagent for research applications (CAS 539-30-0). It is a colorless to light yellow liquid (C9H12O, 136.19 g/mol) available as a neat liquid, analytical standard, or a 10 mM solution in DMSO for calibration, assay development, and metabolite studies.
  • High listed purity (98.99%) suitable for analytical work.
  • Available as neat liquid, analytical standard, and ready-to-use solution.
  • Suitable for calibration, quantitation, and metabolite research.
  • Liquid: colorless to light yellow; density 0.949 g/cm3.
  • Storage recommendations provided to maintain stability in pure form and in solvent.
Catalog No. 50-003-01817 Supplier Medchemexpress LLC Supplier No. HYW01761310MM/1ML
(Ethoxymethyl)benzene is an endogenous metabolite used as an analytical reference and reagent for research applications (CAS 539-30-0). It is a colorless to light yellow liquid (C9H12O, 136.19 g/mol) available as a neat liquid, analytical standard, or a 10 mM solution in DMSO for calibration, assay development, and metabolite studies.
  • High listed purity (98.99%) suitable for analytical work.
  • Available as neat liquid, analytical standard, and ready-to-use solution.
  • Suitable for calibration, quantitation, and metabolite research.
  • Liquid: colorless to light yellow; density 0.949 g/cm3.
  • Storage recommendations provided to maintain stability in pure form and in solvent.