Learn More
Crescent Chemical Co Inc ETHOXYQUIN 250 MG
SDP
Catalog No. 5077699
Encompass_Preferred
Ethoxyquin Fungicide (WS) CAS 91-53-2 Molecular Weight - 217.3068 Storage - -18°C Catalog Num - DRE-CA13310000 pesticide
Catalog No. 50-776-99 Supplier Crescent Chemical Co Inc Supplier No. PSCA13310000
Ethoxyquin Fungicide (WS) CAS 91-53-2 Molecular Weight - 217.3068 Storage - -18°C Catalog Num - DRE-CA13310000 pesticide