Learn More
Medchemexpress LLC Ethoxyquin | 91-53-2 | 98.8% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB142510MM/1ML
Encompass_Preferred
Ethoxyquin is an antioxidant commonly used in animal feed and acts as an inhibitor of heat shock protein 90 (Hsp90). It has been shown to provide neuroprotection in studies, with optimal efficacy in the nanomolar range. Ethoxyquin does not interfere with Paclitaxel's ability to induce cell death in breast cancer cell lines.
  • Antioxidant properties
  • Hsp90 inhibitor
  • Provides neuroprotection at specific concentrations
  • Does not block Paclitaxel-induced cell death
Catalog No. 50-004-12706