Learn More
Medchemexpress LLC Ethoxyquin | 91-53-2 | 98.8% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB14255G
Encompass_Preferred
Ethoxyquin is an antioxidant that has been utilized in animal feed for numerous years, and it also functions as an inhibitor of heat shock protein 90 (Hsp90).
  • Antioxidant
  • Inhibitor of heat shock protein 90 (Hsp90)
  • Provides neuroprotection against chemotherapy-induced neurotoxicity via Hsp90 modulation
  • Provides neuroprotection against cisplatin-induced neurotoxicity
  • Exhibits dose-dependent neuroprotection
Catalog No. 50-004-13551